| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000412376.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
ENST00000613141.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900010123.hg.1 |
NR_002836.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012140.hg.1 |
ENST00000587852.2 |
Ensembl |