| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000379404.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000404933.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000457085.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000457085.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000457085.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000457085.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000457085.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000457085.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000457085.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000478094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000478094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000478094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000478094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000478094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
ENST00000478094.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_001258423.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00006770.hg.1 |
NM_004595.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012254.hg.1 |
ENST00000454078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012254.hg.1 |
ENST00000454078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012254.hg.1 |
ENST00000454078.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600012254.hg.1 |
ENST00000454078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012254.hg.1 |
ENST00000454078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012254.hg.1 |
ENST00000454078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012254.hg.1 |
ENST00000454078.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600012254.hg.1 |
ENST00000454078.1 |
Ensembl |