Search
Search

CXorf21| Gene Symbol: CXorf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025159.2 | NP_079435.1 | 3 | 914 | 99 | |
| GenBank mRNA | AK021639.1 | - | 3 | 795 | 99 | |
| BC020611.1 | - | 3 | 842 | 4768735 | 99 | |
| BX281759.1 | - | 1 | 374 | 99 | ||
Set Membership: |
|