Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs04999718_s1
          See other SLC10A3 GE Assays ›
          Gene Symbol:
          SLC10A3
          Gene Name:
          solute carrier family 10 member 3
          Gene Aliases:
          DXS253E, P3
          Chromosome Location:
          Chr.X: 154487306 - 154490690 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs04999718_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SLC10A3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001142392.2 NP_001135864.1 3 712 110
          NM_019848.4 NP_062822.1 2 947 110
          XM_005277913.4 XP_005277970.1 2 1369 110
          XM_006724847.3 XP_006724910.1 2 890 110
          XM_011531200.2 XP_011529502.2 2 922 110
          XM_011531201.2 XP_011529503.1 2 626 110
          GenBank mRNA BX401493.2 - 3 684 110

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SLC10A3

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          bile acid and bile salt transport
          response to retinoic acid
          transmembrane transport
          bile acid:sodium symporter activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000263512.5 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000369649.8 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393586.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000393587.4 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 ENST00000453912.1 Ensembl
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142391.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_001142392.2 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq
          Affymetrix Human Clariom D Assay TC0X00011206.hg.1 NM_019848.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0