| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000315289.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000373789.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
ENST00000395260.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_001282941.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000010758.hg.1 |
NM_145307.3 |
Refseq |