Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05001569_m1
          See other CFAP58 GE Assays ›
          Gene Symbol:
          CFAP58
          Gene Name:
          cilia and flagella associated protein 58
          Gene Aliases:
          C10orf80, CCDC147, bA127L20.4, bA127L20.5, bA554P13.1
          Chromosome Location:
          Chr.10: 104339124 - 104455090 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05001569_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CFAP58
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008723.1 NP_001008723.1 6 - 7 1070 74
          XM_011539377.2 XP_011537679.1 6 - 7 1789 74
          XM_017015787.1 XP_016871276.1 6 - 7 1441 74
          XM_017015788.1 XP_016871277.1 6 - 7 1972 74
          XM_017015789.1 XP_016871278.1 6 - 7 1059 74
          XM_017015790.1 XP_016871279.1 6 - 7 1059 74
          GenBank mRNA AK093227.1 - 6 - 7 1058 74
          AK296463.1 - 6 - 7 1063 74
          BC036225.1 - 6 - 7 1072 5272786 74
          BC109126.2 - 6 - 7 1024 40036085 74
          BX641120.1 - 6 - 7 1069 74

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CFAP58

          Panther Classification:

          Molecular Function -

          structural protein

          Gene Ontology Categories:

          Process(es)

          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369703.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369703.1 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 ENST00000369704.7 Ensembl
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq
          Affymetrix Human Clariom D Assay TC1000008806.hg.1 NM_001008723.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0