| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000216520.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000216520.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000216520.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000216520.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000555373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000555373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000555373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000557697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
ENST00000557697.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000216520.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000216520.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000216520.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000216520.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000555373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000555373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000555373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000557016.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000557697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
ENST00000557697.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009548.hg.1 |
NM_004450.2 |
Refseq |