| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000357461.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554278.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000554480.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000555994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557006.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
ENST00000557779.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010763.hg.1 |
NM_182526.2 |
Refseq |