| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000585457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000588785.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000591166.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000592897.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
ENST00000618557.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049729.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1700010814.hg.1 |
NR_049730.1 |
Refseq |