| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007239.hg.1 |
ENST00000621775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007239.hg.1 |
ENST00000621775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007239.hg.1 |
ENST00000621775.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007239.hg.1 |
NM_001145073.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007239.hg.1 |
NM_001145073.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007239.hg.1 |
NM_001145073.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00007239.hg.1 |
NM_001145073.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007239.hg.1 |
ENST00000621775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007239.hg.1 |
ENST00000621775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007239.hg.1 |
ENST00000621775.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00007239.hg.1 |
NM_001145073.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007239.hg.1 |
NM_001145073.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007239.hg.1 |
NM_001145073.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00007239.hg.1 |
NM_001145073.2 |
Refseq |