| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000589396.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000592239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000592239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
ENST00000592239.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
NM_152600.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
NM_152600.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
NM_152600.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011483.hg.1 |
NM_152600.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000325421.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000589396.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000592239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000592239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
ENST00000592239.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
NM_152600.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
NM_152600.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
NM_152600.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011483.hg.1 |
NM_152600.2 |
Refseq |