| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000427624.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
ENST00000597582.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NM_138466.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900011620.hg.1 |
NR_049780.1 |
Refseq |