Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05019472_m1
          See other TENT5C-DT GE Assays ›
          Gene Symbol:
          TENT5C-DT
          Gene Name:
          TENT5C divergent transcript
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 117596832 - 117605770 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05019472_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TENT5C-DT
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BU568889.1 - 1 - 2 218 85

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000456126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000456126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000456126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 ENST00000456126.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000425010.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000440801.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000456126.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000456126.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000456126.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 ENST00000456126.1 Ensembl
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 NR_121626.1 Refseq
          Affymetrix Human Clariom D Assay TC0100015424.hg.1 NR_121626.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline