| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011623.hg.1 |
ENST00000443836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011623.hg.1 |
ENST00000443836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011623.hg.1 |
ENST00000443836.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011623.hg.1 |
ENST00000491260.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100011623.hg.1 |
ENST00000491260.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011623.hg.1 |
ENST00000443836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011623.hg.1 |
ENST00000443836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011623.hg.1 |
ENST00000443836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011623.hg.1 |
ENST00000491260.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100011623.hg.1 |
ENST00000491260.2 |
Ensembl |