| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012441.hg.1 |
ENST00000453464.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012441.hg.1 |
ENST00000453464.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012441.hg.1 |
ENST00000453464.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012441.hg.1 |
ENST00000453464.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012441.hg.1 |
NM_001205252.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012441.hg.1 |
NM_001205252.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100012441.hg.1 |
NM_001205252.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012441.hg.1 |
ENST00000453464.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012441.hg.1 |
ENST00000453464.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012441.hg.1 |
ENST00000453464.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012441.hg.1 |
ENST00000453464.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100012441.hg.1 |
NM_001205252.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012441.hg.1 |
NM_001205252.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100012441.hg.1 |
NM_001205252.1 |
Refseq |