| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001135240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001135240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001135240.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100018312.hg.1 |
NM_001135240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000426197.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
ENST00000458626.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001085375.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001135240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001135240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001135240.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100018312.hg.1 |
NM_001135240.1 |
Refseq |