Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05023184_s1
          See other NORAD GE Assays ›
          Gene Symbol:
          NORAD
          Gene Name:
          non-coding RNA activated by DNA damage
          Gene Aliases:
          LINC00657
          Chromosome Location:
          Chr.20: 36045618 - 36050960 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05023184_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: NORAD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_027451.1 1 4823 135

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of mRNA stability
          protein sequestering activity

          MicroArray Probes:

          Agilent Affymetrix ArrayStar Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147256 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31795117 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31795116 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31795062 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147259 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147247 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31795060 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147249 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147257 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147269 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147261 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31795115 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147270 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31795114 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147262 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31795113 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147272 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P31795061 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147245 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P17189408 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147271 - -
          Agilent Human Whole Genome ChIP Tiling Array A_17_P11147246 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 291597 - -
          Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M 472088 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 112685 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 400051 - -
          Agilent SurePrint G3 Human CNV Microarray 2x400K 102348 AK027104.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 102348 BC053733.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 102348 CR594532.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 102348 CR604494.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 102348 CR616776.1 genbank_mrna_human
          Agilent SurePrint G3 Human CNV Microarray 2x400K 102348 NM_024766.2 refseq_mrna_human_NM
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3331726 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3297003 - -
          Agilent Whole Human Genome Oligo Microarray 4x44K 12003 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 31745 - -
          Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray 47349 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3297003 - -
          Agilent Whole Human Genome Microarray 4x44K v2 A_33_P3331726 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3331726 - -
          SurePrint G3 Human GE Microarray G4851A A_33_P3297003 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795112 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147271 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795114 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795117 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147262 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147270 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147261 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795112 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147260 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147263 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795116 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P31795115 - -
          Agilent SurePrint G3 Human Promoter Microarray 1x1M A_17_P11147271 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147268 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147264 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147256 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147266 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147270 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147255 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147244 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147248 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P17189459 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147253 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147257 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147272 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147246 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147262 - -
          Agilent Human Whole Genome ChIP-on-Chip A_17_P11147250 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT HG-U133+ PM Array Plate 224597_PM_at - -
          Affymetrix HT HG-U133+ PM Array Plate 1558028_PM_x_at - -
          Affymetrix Human Genome U219 Array 11757398_x_at - -
          Affymetrix Human Genome U219 Array 11757398_PM_x_at - -
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 ENST00000567732.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011138.hg.1 NR_027451.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR20011138.hg.1 TC20000810.hg.1 HTA TC
          Affymetrix Human Transcriptome Array 2.0 TC20000810.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163006.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163006.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163006.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163006.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163006.hg.1 TC2000008995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000163002.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163002.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163002.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163002.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163002.hg.1 TC2000008995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000163003.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163003.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163003.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163003.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163003.hg.1 TC2000008995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000163007.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163007.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163007.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163007.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163007.hg.1 TC2000008995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay TC2000008995.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163004.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163004.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163004.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163004.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163004.hg.1 TC2000008995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR2000163005.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163005.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163005.hg.1 ENST00000565493.1 Ensembl
          Affymetrix Human Clariom D Assay PSR2000163005.hg.1 NR_027451.1 Refseq
          Affymetrix Human Clariom D Assay PSR2000163005.hg.1 TC2000008995.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 ENST00000411553.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 ENST00000567732.1 Ensembl
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 NR_026790.1 Refseq
          Affymetrix Human Clariom D Assay TC0600007310.hg.1 NR_026790.1 Refseq
          Human Genome U95 Set 35603_at - -
          Human Genome U95 Set 35603_PM_at - -
          Human Genome U95 Set 82825_r_at - -
          Human Genome U95 Set 82825_PM_r_at - -
          Human Genome U95 Set 69049_f_at - -
          Human Genome U95 Set 69049_PM_f_at - -
          Human Genome U95 Set 72322_at - -
          Human Genome U95 Set 72322_PM_at - -
          Array Name Probe IDs Targets Target Type
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A017624 - -
          Arraystar Human LncRNA microarray V2.0 ASHG19A3A017624 - -
          Array Name Probe IDs Targets Target Type
          HumanHT-12 v4 Expression BeadChip ILMN_1655026 NR_027451.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1655026 AK090641.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1655026 BC011592.1 human_transcriptome
          HumanHT-12 v4 Expression BeadChip ILMN_1655026 CR612021.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655026 NR_027451.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655026 AK090641.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655026 BC011592.1 human_transcriptome
          Illumina HumanWG-6 v2.0 expression beadchip ILMN_1655026 CR612021.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655026 NR_027451.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655026 AK090641.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655026 BC011592.1 human_transcriptome
          Illumina human-6 v2.0 expression beadchip ILMN_1655026 CR612021.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655026 NR_027451.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655026 AK090641.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655026 BC011592.1 human_transcriptome
          Illumina HumanWG-6 v3.0 expression beadchip ILMN_1655026 CR612021.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1655026 NR_027451.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1655026 AK090641.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1655026 BC011592.1 human_transcriptome
          Illumina HumanHT-12 V3.0 expression beadchip ILMN_1655026 CR612021.1 human_transcriptome

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline