Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05025402_g1
          See other OOSP1 GE Assays ›
          Gene Symbol:
          OOSP1
          Gene Name:
          oocyte secreted protein 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 26065 - 47135 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05025402_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: OOSP1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001395276.1 NP_001382205.1 2 - 3 287 118
          XM_047426711.1 XP_047282667.1 3 - 4 324 118
          GenBank mRNA AB231702.1 - 1 - 2 154 118

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Homo sapiens Probe spans exons

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 ENST00000398992.2 Ensembl
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq
          Affymetrix Human Clariom D Assay TC1100007760.hg.1 NR_130719.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline