| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009390.hg.1 |
ENST00000603606.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000415371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000415371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000415371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000415371.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000248924.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000323205.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000415371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000415371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000415371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000415371.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000426858.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000445195.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000451984.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
ENST00000478203.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_001171690.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007319.hg.1 |
NM_014291.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009390.hg.1 |
ENST00000603606.1 |
Ensembl |