| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000425712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000425712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000425712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000425712.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000440711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000440711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000440711.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000425712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000425712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000425712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000425712.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000440711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000440711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000440711.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
ENST00000447933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700013638.hg.1 |
NR_103858.1 |
Refseq |