Search
Search

TTTY19| Gene Symbol: TTTY19 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_001549.1 | 2 | 227 | 70 | |||
| GenBank mRNA | AF527831.1 | - | LIT2229 TTTY19 |
2 | 227 | 70 | |
Set Membership: |
|