| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000315948.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000453259.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000500869.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
ENST00000509384.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_001286812.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500011435.hg.1 |
NM_198273.2 |
Refseq |