| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371504.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371504.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371504.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371504.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
ENST00000371505.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600011914.hg.1 |
NM_182539.3 |
Refseq |