| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000628266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
ENST00000628266.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000347708.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000525101.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530074.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000530409.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000533266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000534255.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000628266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
ENST00000628266.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0800009242.hg.1 |
NM_016458.3 |
Refseq |