Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05047658_gH
          See other TTPA GE Assays ›
          Gene Symbol:
          TTPA
          Gene Name:
          alpha tocopherol transfer protein
          Gene Aliases:
          ATTP, AVED, TTP1, alphaTTP
          Chromosome Location:
          Chr.8: 63058409 - 63086053 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05047658_gH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TTPA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000370.3 NP_000361.1 1 122 94
          NM_001413414.1 NP_001400343.1 1 122 94
          NM_001413415.1 NP_001400344.1 1 122 94
          NM_001413416.1 NP_001400345.1 1 122 94
          NM_001413417.1 NP_001400346.1 1 122 94
          NM_001413418.1 NP_001400347.1 1 122 94
          NR_182149.1 1 122 94
          NR_182150.1 1 122 94
          NR_182151.1 1 122 94
          GenBank mRNA BC041784.1 - 1 108 94
          BC058000.1 - 1 126 94
          D49488.1 - 1 108 94
          U21938.1 - 1 97 94

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TTPA

          Gene Ontology Categories:

          Function(s) Process(es)

          placenta development
          embryonic placenta development
          lipid metabolic process
          response to toxic substance
          vitamin E metabolic process
          vitamin transport
          negative regulation of establishment of blood-brain barrier
          intermembrane lipid transfer
          positive regulation of beta-amyloid clearance
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          lipid binding
          vitamin E binding
          phosphatidylinositol-3,4-bisphosphate binding
          intermembrane lipid transfer activity
          phosphatidylinositol bisphosphate binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000521138.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000521138.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 ENST00000521138.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000260116.4 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000521138.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000521138.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 ENST00000521138.1 Ensembl
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq
          Affymetrix Human Clariom D Assay TC0800010631.hg.1 NM_000370.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline