| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522086.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522086.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000517829.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000519819.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521061.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000521221.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0800010638.hg.1 |
ENST00000522265.1 |
Ensembl |