| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000609227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
ENST00000609227.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000463551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000463551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000463551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000463551.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000475354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000475354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
ENST00000475354.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000395539.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000445759.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000446986.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000609227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
ENST00000609227.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010636.hg.1 |
NR_028443.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000317881.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000460013.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000463551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000463551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000463551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000463551.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000475354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000475354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
ENST00000475354.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008249.hg.1 |
NM_001152.4 |
Refseq |