| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000582452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000582452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000582452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000582452.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000584120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000584120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000584120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
ENST00000584120.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578092.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000578613.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000582452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000582452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000582452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000582452.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000584120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000584120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000584120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
ENST00000584120.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1800009250.hg.1 |
NR_015417.1 |
Refseq |