| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000555732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000555732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000555732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000555732.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000554404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000554404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000554404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
ENST00000557719.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554538.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000554742.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000555732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000555732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000555732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000555732.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
ENST00000556290.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400010032.hg.1 |
NR_027004.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000267594.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000554404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000554404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000554404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000556222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557000.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
ENST00000557719.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207071.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207072.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207073.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_001207074.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400008048.hg.1 |
NM_138344.4 |
Refseq |