| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
ENST00000448599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
ENST00000448599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
ENST00000448599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
ENST00000448599.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
ENST00000560745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
ENST00000560745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
ENST00000560745.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
NM_001145643.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
NM_001145643.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
NM_001145643.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500006948.hg.1 |
NM_001145643.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
ENST00000448599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
ENST00000448599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
ENST00000448599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
ENST00000448599.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
ENST00000560745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
ENST00000560745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
ENST00000560745.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
NM_001145643.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
NM_001145643.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
NM_001145643.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500006948.hg.1 |
NM_001145643.1 |
Refseq |