| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000598726.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000307468.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000536878.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597219.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000597850.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598689.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000598726.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000599456.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
ENST00000600053.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900009001.hg.1 |
NM_001010879.3 |
Refseq |