| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000385267.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600016.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000601711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000601711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
ENST00000601711.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900008691.hg.1 |
NR_030373.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000385267.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000439461.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593612.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000593703.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595000.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000595149.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000599581.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600016.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000600821.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000601711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000601711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
ENST00000601711.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NM_001010851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900008691.hg.1 |
NR_030373.1 |
Refseq |