Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05053086_s1
          See other SIX5 GE Assays ›
          Gene Symbol:
          SIX5
          Gene Name:
          SIX homeobox 5
          Gene Aliases:
          BOR2, DMAHP
          Chromosome Location:
          Chr.19: 45764785 - 45769239 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05053086_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: SIX5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175875.4 NP_787071.2 1 731 65
          GenBank mRNA AL540911.3 - 1 65
          BU859227.1 - 145 65
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CR622406.1 - 1 Chr. 19 - 45768387 - 45769685 [+] on Build GRCh38 65

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene SIX5

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          lens development in camera-type eye
          transcription, DNA-templated
          spermatid development
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of skeletal muscle satellite cell proliferation
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000559756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000559756.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586498.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586498.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000586498.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000317578.6 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560160.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000560168.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 ENST00000622857.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900010953.hg.1 NM_175875.4 Refseq
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000559756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000559756.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586251.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586498.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586498.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000586498.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000590076.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000591530.1 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000592217.2 Ensembl
          Affymetrix Human Clariom D Assay TC1900008347.hg.1 ENST00000592217.2 Ensembl

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline