| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |