Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05057081_s1
          See other MGME1 GE Assays ›
          Gene Symbol:
          MGME1
          Gene Name:
          mitochondrial genome maintenance exonuclease 1
          Gene Aliases:
          C20orf72, DDK1, MTDPS11, bA504H3.4
          Chromosome Location:
          Chr.20: 17968594 - 17991122 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05057081_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: MGME1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001310338.1 NP_001297267.1 2 375 86
          NM_001310339.1 NP_001297268.1 2 375 86
          NM_052865.3 NP_443097.1 2 375 86
          XM_005260867.2 XP_005260924.1 2 597 86
          XM_005260870.3 XP_005260927.1 2 323 86
          XM_006723663.3 XP_006723726.1 2 323 86
          XM_011529394.1 XP_011527696.1 2 212 86
          XM_011529395.1 XP_011527697.1 2 375 86
          XM_017028127.1 XP_016883616.1 2 597 86
          XM_017028128.1 XP_016883617.1 2 471 86
          GenBank mRNA AF306684.1 - 2 183 86
          AF306685.1 - 2 183 86
          AF306686.1 - 2 221 86
          AK027503.1 - 2 179 86
          AK315532.1 - 2 211 86
          BC016869.1 - 2 147 3850853 86
          BX382492.2 - 1 140 86
          CV807140.1 - 2 188 86
          DB014120.1 - 2 375 86

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene MGME1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial genome maintenance
          mitochondrial DNA replication
          mitochondrial DNA repair
          nucleic acid phosphodiester bond hydrolysis
          single-stranded DNA exodeoxyribonuclease activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377704.4 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377709.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000377710.9 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000463219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000463219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000463219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000463219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000463219.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000467391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000467391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000467391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000467391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 ENST00000467391.1 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310338.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_001310339.1 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC2000006813.hg.1 NM_052865.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-tlt54:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0