| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000609782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000609782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000609782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000609782.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000631186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000631186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000631186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
ENST00000631186.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017924.hg.1 |
ENST00000455599.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100017924.hg.1 |
ENST00000455599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017924.hg.1 |
ENST00000455599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100017924.hg.1 |
ENST00000455599.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000423808.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000426534.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000436878.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000458316.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000609782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000609782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000609782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000609782.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000616647.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000631186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000631186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000631186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
ENST00000631186.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126010.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2100006834.hg.1 |
NR_126011.1 |
Refseq |