Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs05502358_s1
          See other HOTAIR GE Assays ›
          Gene Symbol:
          HOTAIR
          Gene Name:
          HOX transcript antisense RNA
          Gene Aliases:
          HOXAS, HOXC-AS4, HOXC11-AS1, NCRNA00072
          Chromosome Location:
          Chr.12: 53962312 - 53974954 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs05502358_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HOTAIR
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003716.4 4 359 80
          NR_047517.2 5 501 80
          NR_047518.2 3 229 80
          NR_186237.1 4 359 80
          NR_186238.1 5 440 80
          NR_186239.1 4 331 80
          NR_186240.1 6 625 80
          NR_186241.1 5 501 80
          NR_186242.1 4 347 80
          NR_186243.1 5 473 80
          NR_186244.1 5 473 80
          NR_186245.1 4 371 80
          NR_186246.1 6 597 80
          NR_186247.1 4 375 80
          NR_186248.1 5 501 80
          NR_186249.1 5 497 80
          NR_186250.1 5 501 80
          GenBank mRNA AK123741.1 - 3 257 80
          DA567507.1 - 4 419 80
          DA905601.1 - 5 503 80
          DA907429.1 - 3 473 80
          DQ926657.1 - 4 364 80
          HG975398.1 - 4 419 80
          MF999225.1 - 2 180 80
          OK349298.1 - 4 400 80
          OK349299.1 - 4 400 80
          OK349300.1 - 3 253 80
          OK349301.1 - 4 320 80
          OK349302.1 - 3 283 80
          OK349303.1 - 3 225 80
          OK349304.1 - 3 248 80
          OK349305.1 - 4 353 80
          OK349306.1 - 3 231 80
          OK349307.1 - 3 232 80
          OK349308.1 - 3 289 80
          OK349309.1 - 4 400 80
          OK349310.1 - 3 274 80
          OK349311.1 - 2 475 80
          OK349312.1 - 3 248 80
          OK349313.1 - 5 440 80
          OK349314.1 - 2 240 80
          OK349315.1 - 2 79 80
          OK349316.1 - 5 755 80
          OK349317.1 - 3 178 80
          OK349318.1 - 1 50 80
          OK349319.1 - 5 560 80
          OK349320.1 - 6 832 80
          OK349321.1 - 5 507 80
          OK349322.1 - 1 181 80
          OK349323.1 - 5 541 80
          OK349324.1 - 6 705 80
          OK349325.1 - 4 499 80
          OK349326.1 - 6 619 80
          OK349327.1 - 5 640 80
          OK349328.1 - 4 359 80
          OK349329.1 - 5 440 80
          OK349330.1 - 6 951 80
          OK349331.1 - 5 476 80

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline