Search
Search

SURF4| Gene Symbol: SURF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001280788.1 | NP_001267717.1 | 6 | 1575 | 98 | |
| NM_001280789.1 | NP_001267718.1 | 6 | 1361 | 98 | ||
| NM_001280790.1 | NP_001267719.1 | 7 | 1870 | 98 | ||
| NM_001280791.1 | NP_001267720.1 | 6 | 1818 | 98 | ||
| NM_001280792.1 | NP_001267721.1 | 5 | 1533 | 98 | ||
| NM_001280793.1 | NP_001267722.1 | 5 | 1643 | 98 | ||
| NM_033161.3 | NP_149351.1 | 6 | 1720 | 98 | ||
| XM_011518943.1 | XP_011517245.1 | 8 | 2132 | 98 | ||
| XM_011518944.2 | XP_011517246.1 | 9 | 2066 | 98 | ||
| XM_011518945.2 | XP_011517247.1 | 5 | 3708 | 98 | ||
| XM_017015062.1 | XP_016870551.1 | 8 | 1955 | 98 | ||
| GenBank mRNA | AK026646.1 | - | 1 | 409 | 98 | |
| BX647961.1 | - | 6 | 1573 | 98 | ||
Set Membership: |
|