Search
Search

ZNF589| Gene Symbol: ZNF589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016089.2 | NP_057173.2 | 4 | 1331 | 80 | |
| XM_017006575.1 | XP_016862064.1 | 4 | 1412 | 80 | ||
| XM_017006576.1 | XP_016862065.1 | 6 | 1561 | 80 | ||
| XM_017006577.1 | XP_016862066.1 | 4 | 2617 | 80 | ||
| XM_017006578.1 | XP_016862067.1 | 4 | 1392 | 80 | ||
| XM_017006579.1 | XP_016862068.1 | 6 | 1517 | 80 | ||
| GenBank mRNA | AB209387.1 | - | 4 | 1312 | 80 | |
| AF114817.1 | - | 6 | 1634 | 80 | ||
| AL832831.1 | - | 4 | 1316 | 80 | ||
| AY258146.1 | - | 4 | 1331 | 80 | ||
| BC005859.2 | - | 4 | 1290 | 80 | ||
| BC006247.2 | - | 4 | 1311 | 80 | ||
| BC048798.1 | - | 4 | 1308 | 80 | ||
Set Membership: |
|