Search
Search
| Gene Symbol: RLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001311197.1 | NP_001298126.1 | 3 | 989 | 65 | |
| NM_080864.3 | NP_543140.1 | 2 | 877 | 65 | ||
| GenBank mRNA | AY358181.1 | - | 3 | 1660 | 65 | |
Set Membership: |
|