Search
Search

ZNF578| Gene Symbol: ZNF578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099694.2 | NP_001093164.1 | 6 | 5277 | 94 | |
| NM_001366182.2 | NP_001353111.1 | 6 | 5235 | 94 | ||
| XM_047438189.1 | XP_047294145.1 | 5 | 5175 | 94 | ||
| XM_047438190.1 | XP_047294146.1 | 5 | 5490 | 94 | ||
| XM_047438191.1 | XP_047294147.1 | 5 | 5240 | 94 | ||
| XM_047438192.1 | XP_047294148.1 | 5 | 5346 | 94 | ||
| XM_047438193.1 | XP_047294149.1 | 4 | 5089 | 94 | ||
| XM_047438194.1 | XP_047294150.1 | 4 | 5048 | 94 | ||
Set Membership: |
|