Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs06626790_s1
          See other HTR2A GE Assays ›
          Gene Symbol:
          HTR2A
          Gene Name:
          5-hydroxytryptamine receptor 2A
          Gene Aliases:
          5-HT2A, HTR2
          Chromosome Location:
          Chr.13: 46831546 - 46898082 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs06626790_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: HTR2A
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_000621.5 NP_000612.1 3 1238 66
          NM_001165947.5 NP_001159419.2 2 498 66
          NM_001378924.1 NP_001365853.1 3 989 66
          GenBank mRNA A23337.1 - 2 737 66
          AF498982.1 - 2 530 66
          AK302787.1 - 2 360 66
          AK314132.1 - 3 1018 66
          BC069356.1 - 2 609 66
          BC069576.1 - 2 609 66
          BC074848.2 - 2 557 66
          BC074849.2 - 2 557 66
          BC096839.1 - 2 531 66
          BX647777.1 - 2 305 66
          M86841.1 - 2 506 66
          S71229.1 - 2 580 66
          X57830.1 - 2 675 66

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene HTR2A

          Gene Ontology Categories:

          Function(s) Process(es)

          temperature homeostasis
          positive regulation of cytokine production involved in immune response
          glycolytic process
          cellular calcium ion homeostasis
          signal transduction
          G-protein coupled receptor signaling pathway
          G-protein coupled receptor signaling pathway, coupled to cyclic nucleotide second messenger
          positive regulation of cytosolic calcium ion concentration
          phospholipase C-activating serotonin receptor signaling pathway
          serotonin receptor signaling pathway
          chemical synaptic transmission
          memory
          positive regulation of cell proliferation
          response to xenobiotic stimulus
          positive regulation of phosphatidylinositol biosynthetic process
          regulation of dopamine secretion
          artery smooth muscle contraction
          urinary bladder smooth muscle contraction
          positive regulation of heat generation
          response to cocaine
          negative regulation of potassium ion transport
          positive regulation of neuron apoptotic process
          protein localization to cytoskeleton
          positive regulation of fat cell differentiation
          positive regulation of glycolytic process
          positive regulation of vasoconstriction
          viral entry into host cell
          sensitization
          behavioral response to cocaine
          positive regulation of inflammatory response
          sensory perception of temperature stimulus
          detection of temperature stimulus involved in sensory perception of pain
          detection of mechanical stimulus involved in sensory perception of pain
          release of sequestered calcium ion into cytosol
          negative regulation of synaptic transmission, glutamatergic
          positive regulation of ERK1 and ERK2 cascade
          G protein-coupled serotonin receptor signaling pathway
          presynaptic modulation of chemical synaptic transmission
          positive regulation of execution phase of apoptosis
          positive regulation of platelet aggregation
          positive regulation of DNA biosynthetic process
          Gq/11-coupled serotonin receptor activity
          virus receptor activity
          G-protein coupled receptor activity
          G-protein coupled serotonin receptor activity
          protein binding
          protein tyrosine kinase activator activity
          neurotransmitter receptor activity
          identical protein binding
          macromolecular complex binding
          serotonin binding
          1-(4-iodo-2,5-dimethoxyphenyl)propan-2-amine binding
          serotonin receptor activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline