Search
Search

TMEM167A| Gene Symbol: TMEM167A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174909.5 | NP_777569.1 | 4 | 268 | 113 | |
| GenBank mRNA | BC026285.1 | - | 4 | 217 | 113 | |
| BC028585.1 | - | 4 | 251 | 113 | ||
| BC070122.1 | - | 4 | 245 | 113 | ||
| BC107575.1 | - | 4 | 238 | 113 | ||
| BX647979.1 | - | 4 | 227 | 113 | ||
| DA100343.1 | - | 4 | 370 | 113 | ||
Set Membership: |
|