Search
Search

ZNF440| Gene Symbol: ZNF440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152357.3 | NP_689570.2 | 4 | 1645 | 93 | |
| XM_005259731.5 | XP_005259788.1 | 4 | 1654 | 93 | ||
| XM_017026254.2 | XP_016881743.1 | 3 | 1518 | 93 | ||
| XM_047438145.1 | XP_047294101.1 | 4 | 4361 | 93 | ||
| GenBank mRNA | AK095252.1 | - | 4 | 1626 | 93 | |
| BC035760.1 | - | 4 | 1609 | 93 | ||
Set Membership: |
|