Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs07287153_m1
See other TRIM2 GE Assays ›
Gene Symbol:
TRIM2
Gene Name:
tripartite motif containing 2
Gene Aliases:
CMT2R, RNF86
Chromosome Location:
Chr.4: 153152281 - 153339322 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs07287153_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: TRIM2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001130067.1
NP_001123539.1
2 - 3
336
59
NM_001302692.1
NP_001289621.1
2 - 3
333
59
NM_001302693.1
NP_001289622.1
2 - 3
330
59
NM_001302694.1
NP_001289623.1
2 - 3
435
59
NM_015271.4
NP_056086.2
2 - 3
333
59
XM_006714157.1
XP_006714220.1
2 - 3
333
59
XM_006714158.1
XP_006714221.1
2 - 3
298
59
XM_006714159.3
XP_006714222.1
2 - 3
1144
59
XM_006714160.1
XP_006714223.1
3 - 4
1098
59
XM_006714161.1
XP_006714224.1
3 - 4
1095
59
XM_006714162.1
XP_006714225.1
3 - 4
815
59
XM_011531794.1
XP_011530096.1
2 - 3
412
59
XM_011531795.1
XP_011530097.1
3 - 4
818
59
XM_011531796.2
XP_011530098.1
2 - 3
348
59
XM_011531798.2
XP_011530100.1
2 - 3
453
59
XM_017007943.1
XP_016863432.1
2 - 3
298
59
XM_017007944.1
XP_016863433.1
3 - 4
1098
59
XM_017007945.1
XP_016863434.1
3 - 4
1095
59
XM_017007946.1
XP_016863435.1
2 - 3
1141
59
XM_017007947.1
XP_016863436.1
2 - 3
412
59
XM_017007948.1
XP_016863437.1
2 - 3
366
59
XM_017007949.1
XP_016863438.1
3 - 4
815
59
XM_017007950.1
XP_016863439.1
3 - 4
818
59
XM_017007951.1
XP_016863440.1
2 - 3
512
59
XM_017007952.1
XP_016863441.1
2 - 3
520
59
XM_017007954.1
XP_016863443.1
2 - 3
289
59
XM_017007955.1
XP_016863444.1
2 - 3
282
59
GenBank mRNA
AB011089.1
-
2 - 3
281
59
AF220018.1
-
2 - 3
281
59
AK308731.1
-
2 - 3
301
59
AW071795.1
-
1 - 2
376
59
BC011052.1
-
2 - 3
336
4156234
59
BP310511.1
-
2 - 3
434
59
DA310963.1
-
2 - 3
420
59
DA407606.1
-
2 - 3
298
59
DA631098.1
-
2 - 3
334
59
Back To Top
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017007954.1, XM_017007955.1 transcripts.
Set Membership:
Ambion siRNA available
Amplicon length less than or equal to 70
3' Most
Mammalian Gene Collection
Recommended Assay
Homo sapiens
Probe spans exons
Matching Ambion siRNAs:
Click here to find siRNAs for Gene TRIM2
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination
regulation of neuron apoptotic process
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline