Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07287356_g1
          See other CLTA GE Assays ›
          Gene Symbol:
          CLTA FGFR4
          Gene Name:
          clathrin light chain A
          fibroblast growth factor receptor 4
          Gene Aliases:
          LCA
          CD334, JTK2, TKF
          Chromosome Location:
          Chr.9: 36190874 - 36212059 on Build GRCh38
          Chromosome Location:
          Chr.5: 177086915 - 177098144 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Hs07287356_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: CLTA
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001076677.3 NP_001070145.1 3 - 4 510 69
          NM_001184760.2 NP_001171689.1 3 - 4 510 69
          NM_001184761.2 NP_001171690.1 3 - 4 510 69
          NM_001311203.2 NP_001298132.1 3 - 4 237 69
          NM_001311206.2 NP_001298135.1 2 - 3 472 69
          NM_001833.4 NP_001824.1 3 - 4 510 69
          NM_007096.4 NP_009027.1 3 - 4 510 69
          XM_017014257.2 XP_016869746.1 3 - 4 195 69
          XM_017014258.2 XP_016869747.1 3 - 4 195 69
          GenBank mRNA AK225153.1 - 3 - 4 468 69
          AK291078.1 - 3 - 4 515 69
          AK295692.1 - 3 - 4 499 69
          BC009201.2 - 3 - 4 494 69
          BC019287.1 - 3 - 4 463 69
          BF797444.1 - 2 - 3 425 69
          BM423666.1 - 3 - 4 439 69
          BT007170.1 - 3 - 4 374 69
          CN300759.1 - 3 - 4 220 69
          DQ270158.1 - 3 - 4 374 69
          M20471.1 - 3 - 4 448 69
          M20472.1 - 3 - 4 456 69
          Gene Symbol: FGFR4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA JN007480.1 - 3 - 4 494 69

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001311206.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene CLTA
          Click here to find siRNAs for Gene FGFR4

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          endocytosis
          vesicle-mediated transport
          clathrin coat assembly
          synaptic vesicle endocytosis
          cell division
          clathrin-mediated endocytosis
          structural molecule activity
          protein binding
          clathrin heavy chain binding
          peptide binding
          macromolecular complex binding
          GTPase binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline