Search
Search

TMEM230| Gene Symbol: TMEM230 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009923.1 | NP_001009923.1 | 3 - 4 | 342 | 107 | |
| NM_001009924.1 | NP_001009924.1 | 3 - 4 | 560 | 107 | ||
| NM_001009925.1 | NP_001009925.1 | 2 - 3 | 236 | 107 | ||
| NM_014145.4 | NP_054864.3 | 2 - 3 | 467 | 107 | ||
| XM_005260713.3 | XP_005260770.1 | 2 - 3 | 434 | 107 | ||
| XM_011529227.2 | XP_011527529.1 | 2 - 3 | 457 | 107 | ||
| XM_011529229.2 | XP_011527531.1 | 3 - 4 | 482 | 107 | ||
| XM_017027834.1 | XP_016883323.1 | 2 - 3 | 215 | 107 | ||
| XM_017027835.1 | XP_016883324.1 | 2 - 3 | 236 | 107 | ||
| XM_017027836.1 | XP_016883325.1 | 2 - 3 | 219 | 107 | ||
| GenBank mRNA | AF161392.1 | - | 3 - 4 | 325 | 107 | |
| AK055700.1 | - | 3 - 4 | 516 | 107 | ||
| AK309823.1 | - | 2 - 3 | 236 | 107 | ||
| AK315606.1 | - | 2 - 3 | 226 | 107 | ||
| AY359115.1 | - | 2 - 3 | 359 | 107 | ||
| BC009768.1 | - | 2 - 3 | 220 | 4092724 | 107 | |
| BC009769.1 | - | 2 - 3 | 226 | 4132400 | 107 | |
| BC009770.1 | - | 2 - 3 | 220 | 4249231 | 107 | |
| BC011990.1 | - | 2 - 3 | 207 | 3857616 | 107 | |
| BC015113.1 | - | 2 - 3 | 171 | 4049350 | 107 | |
| BC070212.1 | - | 2 - 3 | 236 | 4687557 | 107 | |
| BC110408.1 | - | 2 - 3 | 198 | 5740520 | 107 | |
Set Membership: |
|