Search
Search

CFHR1| Gene Symbol: CFHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002113.2 | NP_002104.2 | 3 - 4 | 545 | 90 | |
| XM_011509457.2 | XP_011507759.1 | 2 - 3 | 351 | 90 | ||
| GenBank mRNA | BC016755.1 | - | 3 - 4 | 523 | 3934474 | 90 |
| BC107771.1 | - | 3 - 4 | 541 | 4712577 | 90 | |
| BG568702.1 | - | 3 - 4 | 543 | 89 | ||
| M65293.1 | - | 3 - 4 | 506 | 90 | ||
| X56210.1 | - | 3 - 4 | 471 | 90 | ||
Set Membership: |
|