Search
Search

LCN10| Gene Symbol: LCN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001712.3 | NP_001001712.2 | 3 - 4 | 410 | 84 | |
| GenBank mRNA | BC040963.1 | - | 3 - 4 | 1674 | 84 | |
| BC133045.1 | - | 3 - 4 | 429 | 84 | ||
Set Membership: |
|