Search
Search

TMEM258| Gene Symbol: TMEM258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014206.4 | NP_055021.1 | 2 - 3 | 154 | 106 | |
| GenBank mRNA | AF070661.1 | - | 2 - 3 | 168 | 106 | |
| AF085986.1 | - | 2 - 3 | 141 | 106 | ||
| AF086763.1 | - | 2 - 3 | 164 | 106 | ||
| AK291683.1 | - | 2 - 3 | 165 | 106 | ||
| BC002750.2 | - | 2 - 3 | 139 | 106 | ||
| BC015968.1 | - | 2 - 3 | 169 | 106 | ||
Set Membership: |
|